missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl (S)-1,2,3,4-tetrahydroisoquinoline-3-carboxylate hydrochloride, 98%

Catalog No. p-7055798
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH64468MD 250 mg
AAH6446803 1 g
AAH6446806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH64468MD Supplier Thermo Scientific Chemicals Supplier No. H64468MD

CAS: 77497-74-6 | C14H19NO2 | 233.311 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.

Chemical Identifiers

CAS 77497-74-6
Molecular Formula C14H19NO2
Molecular Weight (g/mol) 233.311
MDL Number MFCD04114841
InChI Key KRSUDBIZXKOKGA-LBPRGKRZSA-N
Synonym s-tert-butyl 1,2,3,4-tetrahydroisoquinoline-3-carboxylate, s-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid t-butyl ester, tert-butyl 3s-1,2,3,4-tetrahydroisoquinoline-3-carboxylate, tert-butyl s-1,2,3,4-tetrahydro-isoquinoline-3-carboxylate, s-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid tert-butyl ester, tic-obut, s-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic ac, tert-butyl s-1,2,3,4-tetrahydroisoquinoline-3-carboxylate, tert.-butyl 3s-1,2,3,4-tetrahydroisoquinoline-3-carboxylate, s-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid tert-butyl ester hydrochloride
PubChem CID 9209323
IUPAC Name tert-butyl (3S)-1,2,3,4-tetrahydroisoquinoline-3-carboxylate
SMILES CC(C)(C)OC(=O)C1CC2=CC=CC=C2CN1

Specifications

Quantity 250 mg
Beilstein 5744730
Solubility Information Soluble in water.
Formula Weight 269.77
Percent Purity 99%
Physical Form Solid
Chemical Name or Material tert-Butyl (S)-1,2,3,4-tetrahydroisoquinoline-3-carboxylate hydrochloride
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only