missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl Propionate 98.0+%, TCI America™

Catalog No. p-7122877
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B332725G 25 g
B3327100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B332725G Supplier TCI America Supplier No. B332725G

Chemical Identifiers

CAS 20487-40-5
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00009304
InChI Key JAELLLITIZHOGQ-UHFFFAOYSA-N
Synonym tert-butyl propionate, propanoic acid, 1,1-dimethylethyl ester, propionic acid, tert-butyl ester, t-butyl propionate, t-butyl propanoate, tert-butylpropionate, 1,1-dimethylethyl propionate, tert-?butyl propionate, propionic acid tert-butyl ester, acmc-1cr5r
PubChem CID 88561
IUPAC Name tert-butyl propanoate
SMILES CCC(=O)OC(C)(C)C

Specifications

Color Colorless
Boiling Point 119°C
Quantity 25 g
UN Number 1914
Formula Weight 130.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyl Propionate