missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl pivalate, 98%

Catalog No. p-7035672
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5339506 5 g
AAH5339514 25 g
AAH5339522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5339506 Supplier Thermo Scientific Chemicals Supplier No. H5339506

missing translation for 'validMainframeMsg'

CAS: 16474-43-4 | C9H18O2 | 158.24 g/mol

tert-Butyl pivalate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl pivalate is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.

Chemical Identifiers

CAS 16474-43-4
Molecular Formula C9H18O2
Molecular Weight (g/mol) 158.24
MDL Number MFCD01861974
InChI Key VXHFNALHLRWIIU-UHFFFAOYSA-N
PubChem CID 519272
IUPAC Name tert-butyl 2,2-dimethylpropanoate
SMILES CC(C)(C)OC(=O)C(C)(C)C

Specifications

Density 0.813
Boiling Point 134°C to 135°C
Flash Point >62°C (144°F)
Refractive Index 1.392
Linear Formula (CH3)3CCO2C(CH3)3
Quantity 5 g
Synonym tert-butyl pivalate, tert-butyl trimethylacetate, propanoic acid, 2,2-dimethyl, 1,1-dimethylethyl ester, di-tert-butylcarboxylic acid, 2,2-dimethylpropanoic acid tert-butyl ester, 2,2-dimethyl-propionic acid tert-butyl ester, propanoic acid, 2,2-dimethyl-, 1,1-dimethylethyl ester, propanoic acid,2,2-dimethyl-, 1,1-dimethylethyl ester
Formula Weight 158.24
Percent Purity 98%
Chemical Name or Material tert-Butyl pivalate
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only