missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl N-(tert-Butoxycarbonyl)-L-pyroglutamate 98.0+%, TCI America™

Catalog No. p-7124012
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
B52161G 1 g
B52165G 5 g
2 options

Catalog No. B52161G

Supplier: TCI America B52161G

Only null left

Chemical Identifiers

CAS 91229-91-3
Molecular Formula C14H23NO5
Molecular Weight (g/mol) 285.34
MDL Number MFCD03094770
InChI Key INVKHBRFFCQICU-VIFPVBQESA-N
Synonym s-di-tert-butyl 5-oxopyrrolidine-1,2-dicarboxylate, s-n-boc-2-pyrrolidone-5-carboxylic acid tert-butyl ester, s-n-boc-2-pyrrolidone-5-carboxylic acid t-butyl ester, s-n-boc-pyroglutamic acid tert-butyl ester, s-n-alpha-t-butyloxycarbonyl-pyroglutamic acid t-butyl ester, di-tert-butyl s-5-oxopyrrolidine-1,2-dicarboxylate, 1,2-di-tert-butyl 2s-5-oxopyrrolidine-1,2-dicarboxylate, s-n-boc-pyroglutamic acid tertbutyl ester, di-tert-butyl 2s-5-oxopyrrolidine-1,2-dicarboxylate, boc-py
PubChem CID 11129853
IUPAC Name ditert-butyl (2S)-5-oxopyrrolidine-1,2-dicarboxylate
SMILES CC(C)(C)OC(=O)C1CCC(=O)N1C(=O)OC(C)(C)C

Specifications

Melting Point 58°C
Color White
Quantity 1 g
Formula Weight 285.34
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material tert-Butyl N-(tert-Butoxycarbonyl)-L-pyroglutamate