missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl N-Hydroxycarbamate 98.0+%, TCI America™

Catalog No. H08485G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08485G 5 g
H084825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08485G Supplier TCI America Supplier No. H08485G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 36016-38-3
Molecular Formula C5H11NO3
Molecular Weight (g/mol) 133.15
MDL Number MFCD00002107
InChI Key DRDVJQOGFWAVLH-UHFFFAOYSA-N
Synonym n-boc-hydroxylamine, n-tert-butoxycarbonylhydroxylamine, tert-butyl hydroxycarbamate, boc-amino alcohol, carbamic acid, hydroxy-, 1,1-dimethylethyl ester, n-hydroxycarbamic acid tert-butyl ester, tert-butyl-n-hydroxycarbamate, n-tert-boc-hydroxylamine, t-butyl n-hydroxycarbamate, n-tert-butyloxycarbonylhydroxylamine
PubChem CID 97534
IUPAC Name tert-butyl N-hydroxycarbamate
SMILES CC(C)(C)OC(=O)NO

Specifications

Melting Point 57°C
Color White-Yellow
Quantity 5 g
Formula Weight 133.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material tert-Butyl N-Hydroxycarbamate