missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl N-allylcarbamate, 97%

Catalog No. AAH2601409
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2601409 10 g
AAH2601418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2601409 Supplier Thermo Scientific Chemicals Supplier No. H2601409
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 78888-18-3 | C8H15NO2 | 157.213 g/mol

tert-Butyl N-allylcarbamate is used in the synthesis of isoxazolidines. Synthesis of 5-substituted thiazolidin-2-ones is from the reaction of xanthates and tert-butyl N-allylcarbamates .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl N-allylcarbamate is used in the synthesis of isoxazolidines. Synthesis of 5-substituted thiazolidin-2-ones is from the reaction of xanthates and tert-butyl N-allylcarbamates .

Solubility
Insoluble in water. Soluble in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 78888-18-3
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.213
MDL Number MFCD00191870
InChI Key AWARHXCROCWEAK-UHFFFAOYSA-N
Synonym tert-butyl allylcarbamate, tert-butyl n-allylcarbamate, tert-butyl n-prop-2-en-1-yl carbamate, n-boc-allylamine, t-butyl n-allylcarbamate, carbamic acid, 2-propenyl-, 1,1-dimethylethyl ester, acmc-209pfu, allyl tert-butyl carbamate, tert-butyl-n-allylcarbamate, n-tertbutoxycarbonyl allyl amine
PubChem CID 4067274
IUPAC Name tert-butyl N-prop-2-enylcarbamate
SMILES CC(C)(C)OC(=O)NCC=C

Specifications

Melting Point 34°C to 39°C
Boiling Point 48°C to 50°C (1mmHg)
Linear Formula H2C=CHCH2NHCO2C(CH3)3
Solubility Information Insoluble in water. Soluble in methanol.
Density 0.938
Quantity 10 g
Flash Point 79°C (175°F)
Formula Weight 157.21
Percent Purity 97%
Chemical Name or Material tert-Butyl N-allylcarbamate
Hazard Category H227-H315-H319-H335
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only