missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl-N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate, 97+%, Thermo Scientific™

Chemical Identifiers

CAS 330793-01-6
Molecular Formula C17H26BNO4
Molecular Weight (g/mol) 319.21
MDL Number MFCD02179439
InChI Key HSJNIOYPTSKQBD-UHFFFAOYSA-N
Synonym 4-n-boc-amino phenylboronic acid pinacol ester, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 4-boc-amino benzeneboronic acid pinacol ester, tert-butyl n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 4-boc-amino phenylboronic acid pinacol cyclic ester, 4-boc-aminophenylboronic acid, pinacol ester, 4-tert-butoxycarbonyl aminophenylboronic acid, pinacol ester, tert-butyl n-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenylcarbamate
PubChem CID 2734617
IUPAC Name tert-butyl N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)NC(=O)OC(C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC366350010
View Documents Promotion Details
N/A 1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC366350050
View Documents Promotion Details
N/A 5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

330793-01-6
319.21
HSJNIOYPTSKQBD-UHFFFAOYSA-N
2734617
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)NC(=O)OC(C)(C)C
C17H26BNO4
MFCD02179439
4-n-boc-amino phenylboronic acid pinacol ester, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 4-boc-amino benzeneboronic acid pinacol ester, tert-butyl n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 4-boc-amino phenylboronic acid pinacol cyclic ester, 4-boc-aminophenylboronic acid, pinacol ester, 4-tert-butoxycarbonyl aminophenylboronic acid, pinacol ester, tert-butyl n-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenylcarbamate
tert-butyl N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate

Specifications

330793-01-6
97% min. (HPLC)
MFCD02179439
Solubility in water: insoluble in water
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)NC(=O)OC(C)(C)C
319.21
319.21
tert-Butyl-N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate
Authentic
C17H26BNO4
4-n-boc-amino phenylboronic acid pinacol ester, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 4-boc-amino benzeneboronic acid pinacol ester, tert-butyl n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 4-boc-amino phenylboronic acid pinacol cyclic ester, 4-boc-aminophenylboronic acid, pinacol ester, 4-tert-butoxycarbonyl aminophenylboronic acid, pinacol ester, tert-butyl n-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenylcarbamate
HSJNIOYPTSKQBD-UHFFFAOYSA-N
tert-butyl N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate
2734617
97+%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Recommended Storage : Refrigerator +4°C

SDS
Documents

Documents

RUO – Research Use Only