missing translation for 'onlineSavingsMsg'
Learn More

tert.-Butyl methyl malonate, 95%, Thermo Scientific™

Chemical Identifiers

CAS 42726-73-8
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.2
MDL Number MFCD00042856
InChI Key XPSYZCWYRWHVCC-UHFFFAOYSA-N
Synonym tert-butyl methyl malonate, t-butyl methyl malonate, 1-tert-butyl 3-methyl propanedioate, acmc-20ajkv, tert-butyl methylmalote, pubchem20476, methyl tert-butyl malonate, ksc493c8h, tert-butyl methyl malonaoate
PubChem CID 2733872
IUPAC Name 3-O-tert-butyl 1-O-methyl propanedioate
SMILES CC(C)(C)OC(=O)CC(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC339390050
View Documents Promotion Details
N/A 5mL Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC339390250
View Documents Promotion Details
N/A 25mL Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

42726-73-8
174.2
XPSYZCWYRWHVCC-UHFFFAOYSA-N
2733872
CC(C)(C)OC(=O)CC(=O)OC
C8H14O4
MFCD00042856
tert-butyl methyl malonate, t-butyl methyl malonate, 1-tert-butyl 3-methyl propanedioate, acmc-20ajkv, tert-butyl methylmalote, pubchem20476, methyl tert-butyl malonate, ksc493c8h, tert-butyl methyl malonaoate
3-O-tert-butyl 1-O-methyl propanedioate

Specifications

42726-73-8
80°C (11.0mmHg)
Authentic
C8H14O4
H3COOCCH2COOC(CH3)3
1.03
XPSYZCWYRWHVCC-UHFFFAOYSA-N
3-O-tert-butyl 1-O-methyl propanedioate
2733872
95%
1.0300g/mL
79°C
94% min. (GC)
1.4140 to 1.417
MFCD00042856
tert-butyl methyl malonate, t-butyl methyl malonate, 1-tert-butyl 3-methyl propanedioate, acmc-20ajkv, tert-butyl methylmalote, pubchem20476, methyl tert-butyl malonate, ksc493c8h, tert-butyl methyl malonaoate
CC(C)(C)OC(=O)CC(=O)OC
174.2
174.2
tert.-Butyl methyl malonate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 255-919-1

Recommended Storage : Product may discolor to yellow on storage

SDS
Documents

Documents

RUO – Research Use Only