missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl Methyl Ether 99.0+%, TCI America™

Supplier:  TCI America B0991500ML

 View more versions of this product

Catalog No. B0991500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

tert-Butyl Methyl Ether
1634-04-4
55°C
Clear Liquid at 20°C
MFCD00008812
tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane
CC(C)(C)OC
88.15
CHEBI:27642
≥99.0% (GC)
-108°C
Colorless
500 mL
C5H12O
2398
BZLVMXJERCGZMT-UHFFFAOYSA-N
2-methoxy-2-methylpropane
15413
88.15

Chemical Identifiers

1634-04-4
88.15
BZLVMXJERCGZMT-UHFFFAOYSA-N
15413
2-methoxy-2-methylpropane
C5H12O
MFCD00008812
tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane
CHEBI:27642
CC(C)(C)OC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3220

RTECSNumber : KN5250000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions