missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl Methyl Ether 99.0+%, TCI America™

Catalog No. B099125ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B099125ML 25 mL
B0991500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B099125ML Supplier TCI America Supplier No. B099125ML
Only null left

Chemical Identifiers

CAS 1634-04-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00008812
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
Synonym tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane
PubChem CID 15413
ChEBI CHEBI:27642
IUPAC Name 2-methoxy-2-methylpropane
SMILES CC(C)(C)OC

Specifications

Melting Point -108°C
Color Colorless
Quantity 25 mL
Physical Form Clear Liquid at 20°C
UN Number 2398
Boiling Point 55°C
Formula Weight 88.15
Percent Purity ≥99.0% (GC)
Chemical Name or Material tert-Butyl Methyl Ether
EINECSNumber (2)-3220
RTECSNumber KN5250000
TSCA Yes