missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl methyl ether, Honeywell Riedel-de Haën™
Puriss. p.a., ≥99.5% (GC)
$33763.82 - $33763.82
Chemical Identifiers
| CAS | 1634-04-4 |
|---|---|
| Molecular Formula | C5H12O |
| Molecular Weight (g/mol) | 88.15 |
| InChI Key | BZLVMXJERCGZMT-UHFFFAOYSA-N |
| IUPAC Name | 2-methoxy-2-methylpropane |
| SMILES | COC(C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
60-048-590
|
Honeywell Inc
202562027L |
202.7 L | Metal Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.