missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl methyl ether, For pesticide residue analysis, Honeywell Riedel-de Haën™

Catalog No. 6001651
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
4 x 4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001651 2.5 L
6001650 1 L
60046354 4 x 2.5 L
6001653 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 6001651 Supplier Honeywell Chemicals Supplier No. 344982.5L
Only null left

CHROMASOLV™, for pesticide residue analysis

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent

Chemical Identifiers

CAS 1634-04-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00008812
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
Synonym tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane
PubChem CID 15413
ChEBI CHEBI:27642
IUPAC Name 2-methoxy-2-methylpropane
SMILES CC(C)(C)OC

Specifications

Density 0.74g/mL at 25°C
Quantity 2.5 L
Explosion Limit 0.151
Flash Point −33°C (−27.4°F)
Ignition Point 705°F
Linear Formula (CH3)3COCH3
UN Number UN2398
Beilstein 1730942
Formula Weight 88.15g/mol
Percent Purity ≥99.7%
Grade CHROMASOLV
Chemical Name or Material tert-Butyl methyl ether
Show More Show Less
Hazard Statement P210
RTECSNumber KN5250000