missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl methyl ether, For gas chromatography ECD and FID SupraSolv™, MilliporeSigma™
For gas chromatography ECD and FID SupraSolv™
$322.96 - $680.76
Chemical Identifiers
| CAS | 1634-04-4 |
|---|---|
| Molecular Formula | C5H12O |
| Molecular Weight (g/mol) | 88.15 |
| InChI Key | BZLVMXJERCGZMT-UHFFFAOYSA-N |
| Synonym | tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane |
| PubChem CID | 15413 |
| ChEBI | CHEBI:27642 |
| IUPAC Name | 2-methoxy-2-methylpropane |
| SMILES | CC(C)(C)OC |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
M1019951000
|
Technidata
1019951000 |
1 L | Glass Bottle |
Each for $322.96
|
|
||||
|
M1019952500
|
Technidata
1019952500 |
2.5 L | Glass Bottle |
Each for $680.76
|
|
||||
Chemical Identifiers
| 1634-04-4 | |
| 88.15 | |
| tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane | |
| CHEBI:27642 | |
| CC(C)(C)OC |
| C5H12O | |
| BZLVMXJERCGZMT-UHFFFAOYSA-N | |
| 15413 | |
| 2-methoxy-2-methylpropane |
Specifications
| -108.6°C | |
| Colorless | |
| 55.3°C | |
| −28°C (−18.4°F) | |
| Charactristic | |
| Liquid | |
| 268 hPa (20°C) | |
| tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane | |
| BZLVMXJERCGZMT-UHFFFAOYSA-N | |
| 2-methoxy-2-methylpropane | |
| 15413 | |
| 88.15 |
| 1634-04-4 | |
| 0.74g/cm3 at 20°C | |
| 1 L | |
| 460°C (860°F) | |
| Glass Bottle | |
| C5H12O | |
| 2398 | |
| Soluble in water: 42g/L at 20°C | |
| CC(C)(C)OC | |
| 88.15 | |
| CHEBI:27642 | |
| tert-Butyl methyl ether |
Safety and Handling
H315 Causes skin irritation.
H225: Highly flammable liquid and vapour.
Recommended Storage : Keep container tightly closed. Keep container in a cool, well-ventilated place. Keep away from heat and source of ignition. Protected from light.