missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl methyl ether, 99%

Catalog No. AAL14030AU
Change view
Click to view available options
Quantity:
500 mL
1000 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL14030AU 1000 mL
AAL14030AP 500 mL
AAL140300F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL14030AU Supplier Thermo Scientific Chemicals Supplier No. L14030AU

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1634-04-4 | C5H12O | 88.15 g/mol

Used as a chromatography solvent. Acts as a fuel component in fuel for gasoline engines. They raise the oxygen content of gasoline(oxygenates).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a chromatography solvent. Acts as a fuel component in fuel for gasoline engines. They raise the oxygen content of gasoline(oxygenates).

Solubility
Miscible with ethanol and ethyl ether. Immiscible with water.

Notes
Store in a cool, dry, well-ventilated area. It is hygroscopic in nature.

Chemical Identifiers

Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00008812
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
Synonym tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane
PubChem CID 15413
ChEBI CHEBI:27642
IUPAC Name 2-methoxy-2-methylpropane
SMILES CC(C)(C)OC

Specifications

Melting Point -109°C
Density 0.742
Quantity 1000 mL
Flash Point −25°C (−13°F)
Odor Mild
Refractive Index 1.369
UN Number UN2350
Merck Index 146032
Boiling Point 55°C to 56°C
Solubility Information Miscible with ethanol and ethyl ether. Immiscible with water.
Formula Weight 88.15
Percent Purity 0.99
Chemical Name or Material tert-Butyl methyl ether
Show More Show Less
Hazard Category H225-H315-H335
Hazard Statement GHS H Statement
H225-H315
Highly flammable liquid and vapor.
Causes skin irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BUTYL METHYL ETHER
EINECSNumber 216-653-1
RTECSNumber KN5250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only