missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl Methacrylate Monomer (stabilized with MEHQ) 98.0+%, TCI America™
Supplier: TCI America M0326500ML
Specifications
| tert-Butyl Methacrylate Monomer (stabilized with MEHQ) | |
| Colorless | |
| C8H14O2 | |
| 500 mL | |
| Methacrylic Acid tert-Butyl Ester Monomer | |
| CC(=C)C(=O)OC(C)(C)C | |
| 142.198 | |
| 142.20 | |
| Liquid |
| 585-07-9 | |
| 136°C | |
| MFCD00048245 | |
| 3272 | |
| SJMYWORNLPSJQO-UHFFFAOYSA-N | |
| tert-butyl 2-methylprop-2-enoate | |
| 11448 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 585-07-9 | |
| 142.198 | |
| SJMYWORNLPSJQO-UHFFFAOYSA-N | |
| 11448 | |
| CC(=C)C(=O)OC(C)(C)C |
| C8H14O2 | |
| MFCD00048245 | |
| Methacrylic Acid tert-Butyl Ester Monomer | |
| tert-butyl 2-methylprop-2-enoate |
Safety and Handling
EINECSNumber : (2)-1039
RTECSNumber : OZ3675500
TSCA : Yes