missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl Methacrylate Monomer (stabilized with MEHQ) 98.0+%, TCI America™

Catalog No. M032625ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M032625ML 25 mL
M0326500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M032625ML Supplier TCI America Supplier No. M032625ML
Only null left

Chemical Identifiers

CAS 585-07-9
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.198
MDL Number MFCD00048245
InChI Key SJMYWORNLPSJQO-UHFFFAOYSA-N
Synonym Methacrylic Acid tert-Butyl Ester Monomer
PubChem CID 11448
IUPAC Name tert-butyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OC(C)(C)C

Specifications

Color Colorless
Boiling Point 136°C
Quantity 25 mL
UN Number 3272
Formula Weight 142.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyl Methacrylate Monomer (stabilized with MEHQ)
EINECSNumber (2)-1039
RTECSNumber OZ3675500
TSCA Yes