missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl Mercaptan 98.0+%, TCI America™

Catalog No. M0405500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0405500ML 500 mL
M040525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0405500ML Supplier TCI America Supplier No. M0405500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75-66-1
Molecular Formula C4H10S
Molecular Weight (g/mol) 90.184
MDL Number MFCD00004857
InChI Key WMXCDAVJEZZYLT-UHFFFAOYSA-N
Synonym 2-methyl-2-propanethiol, tert-butyl mercaptan, tert-butylthiol, tert-butanethiol, 2-propanethiol, 2-methyl, tert-butylmercaptan, t-butyl mercaptan, t-butylmercaptan, 2-isobutanethiol, tertiary-butyl mercaptan
PubChem CID 6387
IUPAC Name 2-methylpropane-2-thiol
SMILES CC(C)(C)S

Specifications

Melting Point -1°C
Color Colorless
Boiling Point 64°C
Quantity 500 mL
UN Number 2347
Formula Weight 90.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyl Mercaptan
EINECSNumber (2)-0464
RTECSNumber TZ7660000
TSCA Yes