missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl Mercaptan 98.0+%, TCI America™

Catalog No. M040525ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M040525ML 25 mL
M0405500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M040525ML Supplier TCI America Supplier No. M040525ML

Chemical Identifiers

CAS 75-66-1
Molecular Formula C4H10S
Molecular Weight (g/mol) 90.184
MDL Number MFCD00004857
InChI Key WMXCDAVJEZZYLT-UHFFFAOYSA-N
Synonym 2-methyl-2-propanethiol, tert-butyl mercaptan, tert-butylthiol, tert-butanethiol, 2-propanethiol, 2-methyl, tert-butylmercaptan, t-butyl mercaptan, t-butylmercaptan, 2-isobutanethiol, tertiary-butyl mercaptan
PubChem CID 6387
IUPAC Name 2-methylpropane-2-thiol
SMILES CC(C)(C)S

Specifications

Melting Point -1°C
Color Colorless
Boiling Point 64°C
Quantity 25 mL
UN Number 2347
Formula Weight 90.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyl Mercaptan
EINECSNumber (2)-0464
RTECSNumber TZ7660000
TSCA Yes