missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl L-Pyroglutamate 98.0+%, TCI America™

Catalog No. B40261G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40261G 1 g
B40265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40261G Supplier TCI America Supplier No. B40261G

Chemical Identifiers

CAS 35418-16-7
Molecular Formula C9H15NO3
Molecular Weight (g/mol) 185.223
MDL Number MFCD06659481
InChI Key QXGSPAGZWRTTOT-LURJTMIESA-N
Synonym s-tert-butyl 5-oxopyrrolidine-2-carboxylate, s-2-pyrrolidone-5-carboxylic acid t-butyl ester, tert-butyl 5-oxo-l-prolinate, l-pyroglutamic acid tert-butyl ester, tert-butyl s-2-pyrrolidone-5-carboxylate, tert-butyl l-pyroglutamate, tert-butyl 2s-5-oxopyrrolidine-2-carboxylate, s-5-oxo-pyrrolidine-2-carboxylic acid tert-butyl ester, l-pyroglutamicacidtert-butylester, tert-butyl s-5-oxopyrrolidine-2-carboxylate
PubChem CID 10888648
IUPAC Name tert-butyl (2S)-5-oxopyrrolidine-2-carboxylate
SMILES CC(C)(C)OC(=O)C1CCC(=O)N1

Specifications

Melting Point 104°C
Color White
Quantity 1 g
Formula Weight 185.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material tert-Butyl L-Pyroglutamate