missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl Isocyanide 95.0+%, TCI America™

Catalog No. p-7118962
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B12745ML 5 mL
B127425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B12745ML Supplier TCI America Supplier No. B12745ML
Only null left

Chemical Identifiers

CAS 7188-38-7
Molecular Formula C5H9N
Molecular Weight (g/mol) 83.134
MDL Number MFCD00000002
InChI Key FAGLEPBREOXSAC-UHFFFAOYSA-N
Synonym tert-butyl isocyanide, tert-butylisocyanide, t-butylisonitrile, tert-butylisonitrile, tbin, propane, 2-isocyano-2-methyl, 2-methyl-2-isocyanopropane, 2-methylpropyl isocyanide, t-butyl isocyanide, 2-isocyano-2-methyl-propane
PubChem CID 23577
IUPAC Name 2-isocyano-2-methylpropane
SMILES CC(C)(C)[N+]#[C-]

Specifications

Color Colorless
Boiling Point 92°C
Quantity 5 mL
UN Number 1992
Formula Weight 83.13
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyl Isocyanide
RTECSNumber EQ7102500
TSCA No