missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl isocyanide, 98%

Catalog No. AAL1974703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1974703 1 g
AAL1974706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1974703 Supplier Thermo Scientific Chemicals Supplier No. L1974703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7188-38-7 | C5H9N | 83.134 g/mol

tert-Butyl isocyanide is used in a tin-free procedure for alkyl radical reactions in the presence of a free-radical initiator. It is a useful intermediate in multicomponent reactions. It is also used in the synthesis of coumarines, 4H-chromenes, isoxazolines and to trap 2-cyclopropylidene-1,3-diones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl isocyanide is used in a tin-free procedure for alkyl radical reactions in the presence of a free-radical initiator. It is a useful intermediate in multicomponent reactions. It is also used in the synthesis of coumarines, 4H-chromenes, isoxazolines and to trap 2-cyclopropylidene-1,3-diones.

Solubility
Soluble in organic solvents such as ethanol, methanol, ether, toluene and dichloromethane.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 7188-38-7
Molecular Formula C5H9N
Molecular Weight (g/mol) 83.134
MDL Number MFCD00000002
InChI Key FAGLEPBREOXSAC-UHFFFAOYSA-N
Synonym tert-butyl isocyanide, tert-butylisocyanide, t-butylisonitrile, tert-butylisonitrile, tbin, propane, 2-isocyano-2-methyl, 2-methyl-2-isocyanopropane, 2-methylpropyl isocyanide, t-butyl isocyanide, 2-isocyano-2-methyl-propane
PubChem CID 23577
IUPAC Name 2-isocyano-2-methylpropane
SMILES CC(C)(C)[N+]#[C-]

Specifications

Density 0.735
Boiling Point 90°C to 92°C
Flash Point −2°C (28°F)
Odor Stench
Refractive Index 1.3755
Quantity 1 g
UN Number UN1992
Beilstein 1903156
Solubility Information Soluble in organic solvents such as ethanol,methanol,ether,toluene and dichloromethane.
Formula Weight 83.13
Percent Purity 98%
Chemical Name or Material tert-Butyl isocyanide
Show More Show Less
Hazard Category H225-H330
Hazard Statement GHS H Statement
H225-H330
Highly flammable liquid and vapor.
Fatal if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P271-P280-P284-P303+P361+P353-P304+P340-P310-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 230-553-5
RTECSNumber EQ7102500
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only