missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich tert-Butyl hydroperoxide solution
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
$701.38 - $701.38
Specifications
| CAS | 75-91-2 |
|---|---|
| Density | 0.808 g/mL (at 25°C) |
| Molecular Formula | C4H10O2 |
| Linear Formula | (CH3)3 COOH |
| Refractive Index | n20/D 1.402 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
416665100ML
|
Sigma Aldrich
416665100ML |
100 mL |
Each for $701.38
|
|
|||||
Description
tert-Butyl hydroperoxide is an organic peroxide, used in oxidation processes.tert-Butyl hydroperoxide may be used in:osmium catalyzed vicinal hydroxylation of olefins under alkaline conditions catalytic asymmetric oxidation of sulfides to sulfoxides using binaphthol as a chiral auxiliaryoxidation of dibenzothiophenesSpecifications
| 75-91-2 | |
| C4H10O2 | |
| n20/D 1.402 | |
| 1,1-Dimethylethyl hydroperoxide; 2-Hydroperoxy-2-methylpropane; TBHP | |
| 55.4-65.1% (Thiosulphate, titration) |
| 0.808 g/mL (at 25°C) | |
| (CH3)3 COOH | |
| MFCD00002130 | |
| 90.12 |