missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich tert-Butyl hydroperoxide solution

Catalog No. 416665100ML
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
416665100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 416665100ML Supplier Sigma Aldrich Supplier No. 416665100ML
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

tert-Butyl hydroperoxide is an organic peroxide, used in oxidation processes.tert-Butyl hydroperoxide may be used in:osmium catalyzed vicinal hydroxylation of olefins under alkaline conditions catalytic asymmetric oxidation of sulfides to sulfoxides using binaphthol as a chiral auxiliaryoxidation of dibenzothiophenes

Specifications

CAS 75-91-2
Density 0.808 g/mL (at 25°C)
Molecular Formula C4H10O2
Refractive Index n20/D 1.402
Linear Formula (CH3)3 COOH
MDL Number MFCD00002130
Quantity 100 mL
Synonym 1,1-Dimethylethyl hydroperoxide; 2-Hydroperoxy-2-methylpropane; TBHP
Molecular Weight (g/mol) 90.12
Percent Purity 55.4-65.1% (Thiosulphate, titration)