missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl Hydroperoxide, 70% Solution in Water, Spectrum™ Chemical

Catalog No. p-7158568
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18607307 25 mL Amber Glass Bottle
18607308 100 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18607307 Supplier Spectrum Chemical Mfg Cor Supplier No. B249925ML
Only null left

B2499, 75-91-2, C4H10O2

Spectrum™ Chemical tert-Butyl Hydroperoxide, 70 Percent Solution in Water , with a systematic name of tert-Butyl hydroperoxide, is used in a variety of oxidation processes. Spectrum solutions utilize the highest quality raw materials appropriate to your product. Most raw materials meet or exceed the specifications established by the American Chemical Society. Where appropriate, many finished products are traceable to NIST Standard Reference Materials. Manufacturing, quality control testing, and packaging are all performed in Spectrum’s own facilities. Record keeping and sample retention of all produced lots ensure product consistency and complete traceability.

Chemical Identifiers

CAS 75-91-2
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
InChI Key CIHOLLKRGTVIJN-UHFFFAOYSA-N
IUPAC Name 2-methylpropane-2-peroxol
SMILES CC(C)(C)OO

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 25 mL
UN Number UN3109
Grade Reagent