missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl Hydroperoxide, 70% Solution in Water, Spectrum™ Chemical
Description
Spectrum™ Chemical tert-Butyl Hydroperoxide, 70 Percent Solution in Water , with a systematic name of tert-Butyl hydroperoxide, is used in a variety of oxidation processes. Spectrum solutions utilize the highest quality raw materials appropriate to your product. Most raw materials meet or exceed the specifications established by the American Chemical Society. Where appropriate, many finished products are traceable to NIST Standard Reference Materials. Manufacturing, quality control testing, and packaging are all performed in Spectrum’s own facilities. Record keeping and sample retention of all produced lots ensure product consistency and complete traceability.
Specifications
Specifications
| CAS Max % | 1 |
| Boiling Point | 40°C (104°F) |
| Packaging | Amber Glass Bottle |
| Quantity | 25 mL |
| UN Number | UN3109 |
| Grade | Reagent |
| Physical Form | Liquid |
| Chemical Name or Material | tert-Butyl Hydroperoxide, 70 Percent Solution in Water |