missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl hydrogen succinate, 97%

Catalog No. p-7055979
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6470603 1 g
AAH6470606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6470603 Supplier Thermo Scientific Chemicals Supplier No. H6470603

CAS: 15026-17-2 | C8H14O4 | 174.20 g/mol

It is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 15026-17-2
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.20
MDL Number MFCD00273441
InChI Key PCOCFIOYWNCGBM-UHFFFAOYSA-N
Synonym mono-tert-butyl succinate, 4-tert-butoxy-4-oxobutanoic acid, succinic acid mono-tert-butyl ester, tert-butyl hydrogen succinate, mono-t-butyl succinate, pubchem11616, succinic acid 1-tert-butyl ester, succinic acid 4-tert-butyl ester
PubChem CID 4139107
IUPAC Name 4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid
SMILES CC(C)(C)OC(=O)CCC(O)=O

Specifications

Melting Point 51°C to 54°C (literature)
Color Brown-Yellow
Flash Point >110°C (230°F)
Linear Formula HOOCCH2CH2CO2C(CH3)3
Quantity 1 g
Beilstein 1772169
Solubility Information Slightly soluble in water.
Formula Weight 174.2
Percent Purity 97%
Physical Form Solid
Chemical Name or Material tert-Butyl hydrogen succinate
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only