missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl hydrogen malonate, 96%

CAS: 40052-13-9 | C7H12O4 | 160.169 g/mol

$107.33 - $398.96

Chemical Identifiers

CAS 40052-13-9
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.169
MDL Number MFCD00191886
InChI Key NGGGZUAEOKRHMA-UHFFFAOYSA-N
Synonym 3-tert-butoxy-3-oxopropanoic acid, malonic acid mono-tert-butyl ester, mono-tert-butyl malonate, 2-tert-butoxycarbonyl acetic acid, tert-butyl hydrogen malonate, mono-t-butyl malonate, pubchem13754, malonicacidmono-tert-butylester, malonic acid mono t-butyl ester
PubChem CID 545853
IUPAC Name 3-[(2-methylpropan-2-yl)oxy]-3-oxopropanoic acid
SMILES CC(C)(C)OC(=O)CC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2622306
View Documents
Thermo Scientific Chemicals
H2622306
5 g
Each for $107.33
Only null left
 
AAH2622314
View Documents
Thermo Scientific Chemicals
H2622314
25 g
Each for $398.96
Only null left
 
Description

Description

tert-Butyl hydrogen malonate may be used in the preparation of dendritic precursor to asymmetric methanofullerenes, hapten-3,6-(O,S-dimethylthiophosphoramido)-6-oxohexanoic acid, hapten-4,3-(O,S-dimethylthiophosphoramido)-3-oxopropanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl hydrogen malonate may be used in the preparation of dendritic precursor to asymmetric methanofullerenes, hapten-3,6-(O,S-dimethylthiophosphoramido)-6-oxohexanoic acid, hapten-4,3-(O,S-dimethylthiophosphoramido)-3-oxopropanoic acid.

Solubility
Soluble in ethanol.

Notes
Store in a cool, dry condition in a well sealed container.
Specifications

Chemical Identifiers

40052-13-9
160.169
NGGGZUAEOKRHMA-UHFFFAOYSA-N
545853
CC(C)(C)OC(=O)CC(=O)O
C7H12O4
MFCD00191886
3-tert-butoxy-3-oxopropanoic acid, malonic acid mono-tert-butyl ester, mono-tert-butyl malonate, 2-tert-butoxycarbonyl acetic acid, tert-butyl hydrogen malonate, mono-t-butyl malonate, pubchem13754, malonicacidmono-tert-butylester, malonic acid mono t-butyl ester
3-[(2-methylpropan-2-yl)oxy]-3-oxopropanoic acid

Specifications

40052-13-9
1.04
91°C (195°F)
1.426
5 g
3-tert-butoxy-3-oxopropanoic acid, malonic acid mono-tert-butyl ester, mono-tert-butyl malonate, 2-tert-butoxycarbonyl acetic acid, tert-butyl hydrogen malonate, mono-t-butyl malonate, pubchem13754, malonicacidmono-tert-butylester, malonic acid mono t-butyl ester
NGGGZUAEOKRHMA-UHFFFAOYSA-N
3-[(2-methylpropan-2-yl)oxy]-3-oxopropanoic acid
545853
96%
19°C to 20°C
90°C (2 mmHg)
C7H12O4
MFCD00191886
1765457
Soluble in ethanol.
CC(C)(C)OC(=O)CC(=O)O
160.169
160.17
tert-Butyl hydrogen malonate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P235-P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314-H330-H335

DOTInformation : Hazard Class: 8; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only