missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl hydrogen malonate, 96%

Catalog No. AAH2622314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2622314 25 g
AAH2622306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2622314 Supplier Thermo Scientific Chemicals Supplier No. H2622314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 40052-13-9 | C7H12O4 | 160.169 g/mol

tert-Butyl hydrogen malonate may be used in the preparation of dendritic precursor to asymmetric methanofullerenes, hapten-3,6-(O,S-dimethylthiophosphoramido)-6-oxohexanoic acid, hapten-4,3-(O,S-dimethylthiophosphoramido)-3-oxopropanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl hydrogen malonate may be used in the preparation of dendritic precursor to asymmetric methanofullerenes, hapten-3,6-(O,S-dimethylthiophosphoramido)-6-oxohexanoic acid, hapten-4,3-(O,S-dimethylthiophosphoramido)-3-oxopropanoic acid.

Solubility
Soluble in ethanol.

Notes
Store in a cool, dry condition in a well sealed container.

Chemical Identifiers

CAS 40052-13-9
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.169
MDL Number MFCD00191886
InChI Key NGGGZUAEOKRHMA-UHFFFAOYSA-N
Synonym 3-tert-butoxy-3-oxopropanoic acid, malonic acid mono-tert-butyl ester, mono-tert-butyl malonate, 2-tert-butoxycarbonyl acetic acid, tert-butyl hydrogen malonate, mono-t-butyl malonate, pubchem13754, malonicacidmono-tert-butylester, malonic acid mono t-butyl ester
PubChem CID 545853
IUPAC Name 3-[(2-methylpropan-2-yl)oxy]-3-oxopropanoic acid
SMILES CC(C)(C)OC(=O)CC(=O)O

Specifications

Melting Point 19°C to 20°C
Density 1.04
Boiling Point 90°C (2 mmHg)
Flash Point 91°C (195°F)
Refractive Index 1.426
Quantity 25 g
Beilstein 1765457
Solubility Information Soluble in ethanol.
Formula Weight 160.17
Percent Purity 96%
Chemical Name or Material tert-Butyl hydrogen malonate
Show More Show Less
Hazard Category H227-H314-H330-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only