missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl ethyl malonate, 95%

Catalog No. AAA1192714
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1192714 25 g
AAA1192722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1192714 Supplier Thermo Scientific Chemicals Supplier No. A1192714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 32864-38-3 | C9H16O4 | 188.223 g/mol

tert-Butyl ethyl malonate is used in the preparation of polar ester-functionalized aliphatic polysulfone. It is also a useful reagent for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl ethyl malonate is used in the preparation of polar ester-functionalized aliphatic polysulfone. It is also a useful reagent for organic synthesis.

Solubility
Miscible with most common solvents. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 32864-38-3
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.223
MDL Number MFCD00009193
InChI Key OCOBFMZGRJOSOU-UHFFFAOYSA-N
Synonym tert-butyl ethyl malonate, t-butyl ethyl malonate, propanedioic acid, 1,1-dimethylethyl ethyl ester, 1-tert-butyl 3-ethyl propanedioate, tert-butyl ethyl propanedioate, malonic acid tert-butyl ethyl ester, tert-butyl ethyl propane-1,3-dioate, zlchem 692, acmc-1cjju, 1-tert-butyl ethyl malonate
PubChem CID 96345
IUPAC Name 3-O-tert-butyl 1-O-ethyl propanedioate
SMILES CCOC(=O)CC(=O)OC(C)(C)C

Specifications

Density 0.994
Boiling Point 98°C to 100°C (22 mmHg)
Flash Point 87°C (188°F)
Refractive Index 1.416
Quantity 25 g
Beilstein 1776681
Solubility Information Miscible with most common solvents. Immiscible with water.
Formula Weight 188.22
Percent Purity 95%
Chemical Name or Material tert-Butyl ethyl malonate
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 251-269-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only