missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl Diethylphosphonoacetate 97.0+%, TCI America™

Catalog No. B28145G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28145G 5 g
B28141G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28145G Supplier TCI America Supplier No. B28145G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 27784-76-5
Molecular Formula C10H21O5P
Molecular Weight (g/mol) 252.247
InChI Key NFEGNISFSSLEGU-UHFFFAOYSA-N
Synonym tert-butyl diethylphosphonoacetate, tert-butyl 2-diethoxyphosphoryl acetate, diethyl boc-methyl phosphonate, diethoxy-phosphoryl-acetic acid tert-butyl ester, t-butyl diethylphosphonoacetate, diethyl phosphonoacetic acid tert-butyl ester, t-butyl diethyl phosphonoacetate, diethylphosphonoacetic acid tert-butyl ester, diethyl 2-tert-butoxycarbonylmethyl phosphonate, acetic acid, diethoxyphosphinyl-, 1,1-dimethylethyl ester
PubChem CID 2773685
IUPAC Name tert-butyl 2-diethoxyphosphorylacetate
SMILES CCOP(=O)(CC(=O)OC(C)(C)C)OCC

Specifications

Color Yellow
Boiling Point 152°C
Quantity 5 g
Formula Weight 252.25
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyl Diethylphosphonoacetate