Learn More
tert-Butyl diethylphosphonoacetate, 95%
Description
tert-Butyl diethylphosphonoacetate is used as a reactant for preparation of hydroxymethylated dihydroxyvitamin D3 analogs via Wittig-Horner approach, as potential antitumor agents, synthesis of phosphopeptide mimetic prodrugs targeted to Src homology 2 (SH2) domain of signal transducer and activator of transcription 3 (Stat 3) and bicyclic triaminophosphine-promoted stereoselective synthesis of a,β-unsaturated esters, fluorides, and nitriles from aldehydes and ketones using Wadsworth-Emmons phosphonates
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
tert-Butyl diethylphosphonoacetate is used as a reactant for preparation of hydroxymethylated dihydroxyvitamin D3 analogs via Wittig-Horner approach, as potential antitumor agents, synthesis of phosphopeptide mimetic prodrugs targeted to Src homology 2 (SH2) domain of signal transducer and activator of transcription 3 (Stat 3) and bicyclic triaminophosphine-promoted stereoselective synthesis of a,ß-unsaturated esters, fluorides, and nitriles from aldehydes and ketones using Wadsworth-Emmons phosphonates
Solubility
Not miscible or difficult to mix in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with oxidizing agents.
Specifications
Specifications
| Density | 1.074 |
| Boiling Point | 100°C to 103°C (1.5 mmHg) |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.431 |
| Quantity | 25 g |
| Beilstein | 2050126 |
| Solubility Information | Not miscible or difficult to mix in water. |
| Formula Weight | 252.25 |
| Percent Purity | 95% |
| Chemical Name or Material | tert-Butyl diethylphosphonoacetate |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319 Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 000-000-0 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only