missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl Cyanoacetate 97.0+%, TCI America™

Catalog No. C144125ML
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C144125ML 25 mL
C1441250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C144125ML Supplier TCI America Supplier No. C144125ML

Chemical Identifiers

CAS 1116-98-9
Molecular Formula C7H11NO2
Molecular Weight (g/mol) 141.17
MDL Number MFCD00001938
InChI Key BFNYNEMRWHFIMR-UHFFFAOYSA-N
Synonym tert-butyl cyanoacetate, t-butyl cyanoacetate, acetic acid, cyano-, 1,1-dimethylethyl ester, cyanoacetic acid tert-butyl ester, acetic acid, cyano-, tert-butyl ester, acetic acid, 2-cyano-, 1,1-dimethylethyl ester, tert-butylcyanacetat, t-butylcyanoacetate, tert-butylcyanoacetate, tert butyl cyanoacetate
PubChem CID 70693
IUPAC Name tert-butyl 2-cyanoacetate
SMILES CC(C)(C)OC(=O)CC#N

Specifications

Color Yellow
Boiling Point 108°C
Quantity 25 mL
Formula Weight 141.17
Percent Purity ≥97.0% (GC)
Grade FCC
Physical Form Liquid
Chemical Name or Material tert-Butyl Cyanoacetate
EINECSNumber (2)-1549
TSCA Yes
Recommended Storage Refrigerator