missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl cyanoacetate, 98%

Catalog No. AAL0012322
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0012322 100 g
AAL0012306 5 g
AAL0012314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0012322 Supplier Thermo Scientific Chemicals Supplier No. L0012322

CAS: 1116-98-9 | C7H11NO2 | 141.17 g/mol

tert-Butyl cyanoacetate is used in the synthesis of vinylogous urea. It is also used as a new additive for the sugar nucleoside base coupling step en route to DAPD with improved β-selectivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl cyanoacetate is used in the synthesis of vinylogous urea. It is also used as a new additive for the sugar nucleoside base coupling step en route to DAPD with improved β-selectivity.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 1116-98-9
Molecular Formula C7H11NO2
Molecular Weight (g/mol) 141.17
MDL Number MFCD00001938
InChI Key BFNYNEMRWHFIMR-UHFFFAOYSA-N
Synonym tert-butyl cyanoacetate, t-butyl cyanoacetate, acetic acid, cyano-, 1,1-dimethylethyl ester, cyanoacetic acid tert-butyl ester, acetic acid, cyano-, tert-butyl ester, acetic acid, 2-cyano-, 1,1-dimethylethyl ester, tert-butylcyanacetat, t-butylcyanoacetate, tert-butylcyanoacetate, tert butyl cyanoacetate
PubChem CID 70693
IUPAC Name tert-butyl 2-cyanoacetate
SMILES CC(C)(C)OC(=O)CC#N

Specifications

Density 0.987
Boiling Point 67°C to 68°C (1.5 mmHg)
Flash Point 91°C (195°F)
Odor Odorless
Refractive Index 1.42
Quantity 100 g
Beilstein 1755933
Solubility Information Difficult to mix.
Formula Weight 141.17
Percent Purity 98%
Chemical Name or Material tert-Butyl cyanoacetate
Show More Show Less
Hazard Category H227-H302
Hazard Statement GHS H Statement
H301-H227
Toxic if swallowed.
Combustible liquid.
Precautionary Statement P210-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c
EINECSNumber 214-243-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only