missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl chloride, 98+%

Catalog No. p-7020896
Change view
Click to view available options
Quantity:
1000 mL
250 mL
500 mL
Catalog No. Quantity
AAA13004AK 250 mL
AAA13004AP 500 mL
AAA13004AU 1000 mL
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

Catalog No. AAA13004AK

Supplier: Thermo Scientific Chemicals A13004AK

Only null left
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 507-20-0 | C4H9Cl | 92.566 g/mol

tert-Butyl chloride plays an important role as a starting material to perform nucleophilic substitution reactions in order to prepare alcohol and alkoxides salts. It is used as an alkylating agent for the introduction of tert-butyl group and is also involved in Friedel-Crafts reactions. It is employed as an intermediate for the synthesis of agrochemicals and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl chloride plays an important role as a starting material to perform nucleophilic substitution reactions in order to prepare alcohol and alkoxides salts. It is used as an alkylating agent for the introduction of tert-butyl group and is also involved in Friedel-Crafts reactions. It is employed as an intermediate for the synthesis of agrochemicals and pharmaceuticals.

Solubility
Miscible with alcohol, benzene, chloroform and ether. Slightly miscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 507-20-0
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.566
MDL Number MFCD00000816
InChI Key NBRKLOOSMBRFMH-UHFFFAOYSA-N
Synonym tert-butyl chloride, propane, 2-chloro-2-methyl, tert-butylchloride, chlorotrimethylmethane, trimethylchloromethane, 2-chloroisobutane, t-butyl chloride, t-butylchloride, 2-methyl-2-chloropropane, 2-methyl-2-propyl chloride
PubChem CID 10486
IUPAC Name 2-chloro-2-methylpropane
SMILES CC(C)(C)Cl

Specifications

Melting Point -26°C
Density 0.846
Boiling Point 50°C to 52°C
Flash Point −27°C (−16°F)
Odor Mild
Refractive Index 1.3848
Quantity 250 mL
UN Number UN1127
Beilstein 1730872
Merck Index 14,1562
Solubility Information Miscible with alcohol,benzene,chloroform and ether. Slightly miscible with water.
Formula Weight 92.57
Percent Purity ≥98%
Chemical Name or Material tert-Butyl chloride
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CHLOROBUTANES
EINECSNumber 208-066-4
RTECSNumber TX5040000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only