missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl carbazate, 99%

Catalog No. p-79265
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC107880250 25 g Glass bottle
AC107881000 100 g Glass bottle
AC107885000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC107880250 Supplier Thermo Scientific Chemicals Supplier No. 107880250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 870-46-2 | C5H12N2O2 | 132.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 870-46-2
Molecular Formula C5H12N2O2
Molecular Weight (g/mol) 132.16
MDL Number MFCD00007593
InChI Key DKACXUFSLUYRFU-UHFFFAOYSA-N
Synonym tert-butyl carbazate, tert-butyl hydrazinecarboxylate, t-butyl carbazate, hydrazinecarboxylic acid, 1,1-dimethylethyl ester, t-butylcarbazate, tert-butoxycarbonyl hydrazide, boc-hydrazide, boc-hydrazine, tert-butoxycarbonyl hydrazine, carbazic acid tert-butyl ester
PubChem CID 70091
IUPAC Name tert-butyl N-aminocarbamate
SMILES CC(C)(C)OC(=O)NN

Specifications

Melting Point 37°C to 41°C
Color White to Yellow
Density 1.0200g/mL
Boiling Point 63°C to 65°C (0.1 mmHg)
Flash Point 91°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula H2NNHCO2C(CH3)3
Quantity 25 g
Fieser 01,85; 02,46; 11,88
Specific Gravity 1.02
Solubility Information Solubility in water: .. Other solubilities: soluble in organic solvents
Formula Weight 132.16
Percent Purity 99%
Physical Form Crystalline Lumps
Chemical Name or Material tert-Butyl carbazate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic if swallowed.
Toxic in contact with skin.
Flammable solid.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 212-795-3
TSCA TSCA

RUO – Research Use Only