missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl carbazate, 99%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 37°C to 41°C |
| Color | White to Yellow |
| Density | 1.0200g/mL |
| Boiling Point | 63°C to 65°C (0.1 mmHg) |
| Flash Point | 91°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99% |
| Packaging | Glass bottle |
| Linear Formula | H2NNHCO2C(CH3)3 |
| Quantity | 100 g |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Toxic if swallowed. Toxic in contact with skin. Flammable solid. |
| Precautionary Statement | GHS P Statement: Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 212-795-3 |
| TSCA | TSCA |
RUO – Research Use Only