missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl Carbamate 98.0+%, TCI America™

Catalog No. B31325G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31325G 5 g
B313225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31325G Supplier TCI America Supplier No. B31325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4248-19-5
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.148
MDL Number MFCD00007962
InChI Key LFKDJXLFVYVEFG-UHFFFAOYSA-N
Synonym t-butyl carbamate, carbamic acid tert-butyl ester, boc amine, boc-amide, carbamic acid, 1,1-dimethylethyl ester, boc-nh2, tert-butylcarbamat, n-boc amine, boc-amine, tert butyl carbamate
PubChem CID 77922
IUPAC Name tert-butyl carbamate
SMILES CC(C)(C)OC(=O)N

Specifications

Melting Point 109°C
Color White
Quantity 5 g
Formula Weight 117.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material tert-Butyl Carbamate
EINECSNumber (2)-1712
TSCA No