missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl Carbamate 98.0+%, TCI America™
Supplier: TCI America B313225G
| Quantity | 25 g |
|---|
Chemical Identifiers
| C5H11NO2 | |
| MFCD00007962 | |
| t-butyl carbamate, carbamic acid tert-butyl ester, boc amine, boc-amide, carbamic acid, 1,1-dimethylethyl ester, boc-nh2, tert-butylcarbamat, n-boc amine, boc-amine, tert butyl carbamate | |
| tert-butyl carbamate |
Specifications
| 109°C | |
| 4248-19-5 | |
| C5H11NO2 | |
| t-butyl carbamate, carbamic acid tert-butyl ester, boc amine, boc-amide, carbamic acid, 1,1-dimethylethyl ester, boc-nh2, tert-butylcarbamat, n-boc amine, boc-amine, tert butyl carbamate | |
| CC(C)(C)OC(=O)N | |
| 25 g | |
| 77922 | |
| ≥98.0% (GC) |
| tert-Butyl Carbamate | |
| White | |
| MFCD00007962 | |
| LFKDJXLFVYVEFG-UHFFFAOYSA-N | |
| tert-butyl carbamate | |
| 117.148 | |
| 117.15 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-1712
TSCA : No