missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl carbamate, 98+%

Catalog No. aal0025914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0025914 25 g
AAL0025906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0025914 Supplier Thermo Scientific Chemicals Supplier No. L0025914
Only null left

CAS: 4248-19-5 | C5H11NO2 | 117.148 g/mol

tert-Butyl carbamate is used in palladium-catalyzed synthesis of N-Boc-protected anilines. It was used in the synthesis of tetrasubstituted pyrroles, functionalized with ester or ketone groups at C-3 position.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl carbamate is used in palladium-catalyzed synthesis of N-Boc-protected anilines. It was used in the synthesis of tetrasubstituted pyrroles, functionalized with ester or ketone groups at C-3 position.

Solubility
Soluble in methylene chloride, chloroform, and alcohols. Slightly soluble in petroleum ether and water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

Chemical Identifiers

CAS 4248-19-5
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.148
MDL Number MFCD00007962
InChI Key LFKDJXLFVYVEFG-UHFFFAOYSA-N
Synonym t-butyl carbamate, carbamic acid tert-butyl ester, boc amine, boc-amide, carbamic acid, 1,1-dimethylethyl ester, boc-nh2, tert-butylcarbamat, n-boc amine, boc-amine, tert butyl carbamate
PubChem CID 77922
IUPAC Name tert-butyl carbamate
SMILES CC(C)(C)OC(=O)N

Specifications

Melting Point 106°C to 110°C
Beilstein 1744500
Solubility Information Soluble in methylene chloride,chloroform,and alcohols. Slightly soluble in petroleum ether and water.
Quantity 25 g
Formula Weight 117.15
Percent Purity ≥98%
Chemical Name or Material tert-Butyl carbamate
EINECSNumber 224-209-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only