missing translation for 'onlineSavingsMsg'
Learn More

Tert-Butyl Alcohol (tertiary Butanol; 2-Methyl-2-propanol), Reagents

Catalog No. p-200021128
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337547 1 L
01337546 4 L
01337545 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337547 Supplier Labchem Inc Supplier No. C21202001A
Only null left

Tert-Butyl Alcohol (tertiary Butanol; 2-Methyl-2-propanol), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 75-65-0
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
InChI Key DKGAVHZHDRPRBM-UHFFFAOYSA-N
Synonym 2-Methyl-2-propanol, t-Butanol, tert-Butanol, 2-Methylpropan-2-ol
IUPAC Name 2-methylpropan-2-ol
SMILES CC(C)(C)O

Specifications

Melting Point 25.3°C
Color Colorless
Boiling Point 83°C
Quantity 1 L
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 0.79
Grade Reagent
Chemical Name or Material Tert-Butyl Alcohol (tertiary Butanol, 2-Methyl-2-propanol)