missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl alcohol, anhydrous, 99.5%, packaged under Argon in resealable ChemSeal™ bottles

Catalog No. p-7051726
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
4 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41857AD 50 mL
AA41857AK 250 mL
AA41857K2 1 L
AA41857M1 4 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA41857AD Supplier Thermo Scientific Chemicals Supplier No. 041857AD
Only null left

CAS: 75-65-0 | C4H10O | 74.123 g/mol

tert-Butyl alcohol is used as a solvent and an ethanol denaturant. It is an active ingredient in paint remover. It acts as an intermediate in the production of methyl tertiary butyl ether and ethyl tertiary butyl ether by reaction with methanol and ethanol. It is also used in the preparation of tert-butyl chloride, tert-butyl hydroperoxide, isobutylene and methyl methacrylate. It serves as a fuel additive as well as a gasoline octane booster.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon

Chemical Identifiers

CAS 75-65-0
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00004464
InChI Key DKGAVHZHDRPRBM-UHFFFAOYSA-N
Synonym tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol
PubChem CID 6386
ChEBI CHEBI:45895
IUPAC Name 2-methylpropan-2-ol
SMILES CC(C)(C)O

Specifications

Melting Point 25°C
Color Colorless
Density 0.786 g/mL
Boiling Point 83°C
Flash Point 11°C (52°F)
Assay Percent Range 99.8%
Packaging ChemSeal™ bottles
Refractive Index 1.387
Linear Formula (CH3)3COH
Quantity 50 mL
UN Number UN1120
Beilstein 906698
Merck Index 14,1542
Solubility Information Miscible with water,ethanol,ethyl ether and chloroform.
Formula Weight 74.12
Percent Purity 99.5%
Physical Form Liquid
Chemical Name or Material tert-Butyl alcohol, Anhydrous
Show More Show Less
Hazard Category H225-H319-H332-H335-H336
Hazard Statement GHS H Statement
H225-H332-H319-H335
Highly flammable liquid and vapor.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BUTANOLS
EINECSNumber 200-889-7
RTECSNumber EO1925000
TSCA Yes
storageNote1
  • Packaged under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only