missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl alcohol, ACS, 99+%

CAS: 75-65-0 | C4H10O | 74.123 g/mol

Supplier:  Thermo Scientific Chemicals 033278K2

 View more versions of this product

Catalog No. AA33278K2

Only null left
Explore available promotions

Description

Description

tert-Butyl alcohol is used as a solvent and an ethanol denaturant. It is an active ingredient in paint remover. It acts as an intermediate in the production of methyl tertiary butyl ether and ethyl tertiary butyl ether by reaction with methanol and ethanol. It is also used in the preparation of tert-butyl chloride, tert-butyl hydroperoxide, isobutylene and methyl methacrylate. It serves as a fuel additive as well as a gasoline octane booster.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tert-Butyl alcohol
75-65-0
0.786 g/mL
11°C (52°F)
C4H10O
MFCD00004464
906698
Miscible with water,ethanol,ethyl ether and chloroform.
CC(C)(C)O
74.123
CHEBI:45895
ACS Reagent
For ACS analysis
25°C
83°C
>99+%
1.387
1 L
tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol
DKGAVHZHDRPRBM-UHFFFAOYSA-N
2-methylpropan-2-ol
6386
74.12
Liquid

Chemical Identifiers

75-65-0
74.123
DKGAVHZHDRPRBM-UHFFFAOYSA-N
6386
2-methylpropan-2-ol
C4H10O
MFCD00004464
tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol
CHEBI:45895
CC(C)(C)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c

H225-H319-H332-H335-H336

DOTInformation : Hazard Class: 3; Packaging Group: II

EINECSNumber : 200-889-7

RTECSNumber : EO1925000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only