missing translation for 'onlineSavingsMsg'
Learn More

Tert-Butyl acrylate (stabilisé avec MEHQ) 98,0+%, TCI America™

Catalog No. A1389500ML
Change view
Click to view available options
Quantité:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
A1389500ML 500 mL
A138925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1389500ML Supplier TCI America Supplier No. A1389500ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1663-39-4
Formule moléculaire C7H12O2
Poids moléculaire (g/mol) 128.17
Numéro MDL MFCD00008809
Clé InChI ISXSCDLOGDJUNJ-UHFFFAOYSA-N
Synonyme tert-butyl acrylate, t-butyl acrylate, tert-butyl propenoate, 2-propenoic acid, 1,1-dimethylethyl ester, acrylic acid tert-butyl ester, tert-butylacrylate, acrylic acid, tert-butyl ester, unii-92m2cd1o3g, ccris 7035, 2-propenoic acid tert-butyl ester
PubChem CID 15458
Nom de l’IUPAC Tert-butyl prop-2-énoate
SOURIRES CC(C)(C)OC(=O)C=C

Specifications

Nom ou substance chimique tert-Butyl Acrylate (stabilized with MEHQ)
Couleur Incolore
Point d’ébullition 127 °C
Quantité 500 mL
Numéro UN 2348
Poids de formule 128.17
Pourcentage de pureté ≥98,0% (GC)
Forme physique Liquide
EINECSNumber (2)-0989
TSCA Oui