missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl Acrylate (stabilized with MEHQ) 98.0+%, TCI America™

Catalog No. A138925ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A138925ML 25 mL
A1389500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A138925ML Supplier TCI America Supplier No. A138925ML

Chemical Identifiers

CAS 1663-39-4
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00008809
InChI Key ISXSCDLOGDJUNJ-UHFFFAOYSA-N
Synonym tert-butyl acrylate, t-butyl acrylate, tert-butyl propenoate, 2-propenoic acid, 1,1-dimethylethyl ester, acrylic acid tert-butyl ester, tert-butylacrylate, acrylic acid, tert-butyl ester, unii-92m2cd1o3g, ccris 7035, 2-propenoic acid tert-butyl ester
PubChem CID 15458
IUPAC Name tert-butyl prop-2-enoate
SMILES CC(C)(C)OC(=O)C=C

Specifications

Color Colorless
Boiling Point 127°C
Quantity 25 mL
UN Number 2348
Formula Weight 128.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyl Acrylate (stabilized with MEHQ)
EINECSNumber (2)-0989
TSCA Yes