missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl acrylate, 99%, stab. with 15ppm 4-methoxyphenol

Catalog No. AAL17169AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL17169AE 100 mL
AAL17169AP 500 mL
AAL17169AU 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL17169AE Supplier Thermo Scientific Chemicals Supplier No. L17169AE

CAS: 1663-39-4 | C7H12O2 | 128.17 g/mol

tert-Butyl acrylate is a monomer used in the production of poly(tert-butyl acrylate) polymers, poly(acrylic acid) polyelectrolyte brushes and 3-(3-methyl-but-3-enyloxy)-propionic acid tert-butyl ester. It is also used in paints, coatings, high polymer paper conditioner, nursing polymer products and carbonyl reforming polyolefin. Further, it serves as an important raw material in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl acrylate is a monomer used in the production of poly(tert-butyl acrylate) polymers, poly(acrylic acid) polyelectrolyte brushes and 3-(3-methyl-but-3-enyloxy)-propionic acid tert-butyl ester. It is also used in paints, coatings, high polymer paper conditioner, nursing polymer products and carbonyl reforming polyolefin. Further, it serves as an important raw material in organic synthesis.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 1663-39-4
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00008809
InChI Key ISXSCDLOGDJUNJ-UHFFFAOYSA-N
Synonym tert-butyl acrylate, t-butyl acrylate, tert-butyl propenoate, 2-propenoic acid, 1,1-dimethylethyl ester, acrylic acid tert-butyl ester, tert-butylacrylate, acrylic acid, tert-butyl ester, unii-92m2cd1o3g, ccris 7035, 2-propenoic acid tert-butyl ester
PubChem CID 15458
IUPAC Name tert-butyl prop-2-enoate
SMILES CC(C)(C)OC(=O)C=C

Specifications

Melting Point ∼-69°C
Density 0.883
Boiling Point 121°C
Flash Point 11°C (52°F)
Refractive Index 1.411
Linear Formula CH2=CHCOOC(CH3)3
Quantity 100 mL
UN Number UN3272
Beilstein 1742329
Solubility Information Slightly miscible with water.
Formula Weight 128.17
Percent Purity 99%
Chemical Name or Material tert-Butyl acrylate, stabilized with 15ppm 4-methoxyphenol
Show More Show Less
Hazard Category H225-H302+H312+H332-H315-H317-H319-H335
Hazard Statement GHS H Statement
H225-H302-H312-H332-H315-H317-H335
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P272-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 216-768-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only