missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 5-Norbornene-2-carboxylate (endo- and exo- mixture) 95.0+%, TCI America™

Catalog No. p-7123438
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42675G 5 g
B426725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B42675G Supplier TCI America Supplier No. B42675G

Chemical Identifiers

CAS 154970-45-3
Molecular Formula C12H18O2
Molecular Weight (g/mol) 194.274
MDL Number MFCD18083704
InChI Key BZBMBZJUNPMEBD-UHFFFAOYSA-N
Synonym 5-Norbornene-2-carboxylic Acid tert-Butyl Ester, tert-Butyl Bicyclo[2.2.1]hept-5-ene-2-carboxylate, Bicyclo[2.2.1]hept-5-ene-2-carboxylic Acid tert-Butyl Ester
PubChem CID 15607334
IUPAC Name tert-butyl bicyclo[2.2.1]hept-2-ene-5-carboxylate
SMILES CC(C)(C)OC(=O)C1CC2CC1C=C2

Specifications

Color Yellow
Boiling Point 103°C
Quantity 5 g
Formula Weight 194.27
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyl 5-Norbornene-2-carboxylate (endo- and exo- mixture)