missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylate, 97%, Thermo Scientific™

Catalog No. p-4510406
Change view
Click to view available options
Quantity:
250mg
5g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC53239CB 250mg Amber Glass Bottle
CC53239DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC53239CB Supplier Thermo Scientific Chemicals Supplier No. CC53239CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 864771-44-8
Molecular Formula C18H25BN2O4
Molecular Weight (g/mol) 344.218
InChI Key MMXDKXUKSKMROG-UHFFFAOYSA-N
Synonym tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole-1-carboxylate, 1-boc-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole, tert-butyl 5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole-1-carboxylate, 1-n-boc-indazole-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-indazole-1-carboxylic acid tert-butyl ester, 1-boc-indazole-5-boronic acid pinacol ester, tert-butyl 5-tetramethyl-1,3,2-dioxaborolan-2-yl indazole-1-carboxylate, 1-tert-butoxycarbonyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole, 1h-indazole-1-carboxylic acid, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-, 1,1-dimethylethyl ester, tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indazole-1-carboxylate
PubChem CID 18525862
IUPAC Name tert-butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indazole-1-carboxylate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)N(N=C3)C(=O)OC(C)(C)C

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 344.22
Percent Purity 97%
Chemical Name or Material tert-Butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole-1-carboxylate
Hazard Category Warning