missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylate, 97%, Thermo Scientific™
Specifications
Specifications
| Assay Percent Range | 97% |
| Packaging | Amber Glass Bottle |
| Quantity | 5g |
| Formula Weight | 344.22 |
| Percent Purity | 97% |
| Chemical Name or Material | tert-Butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole-1-carboxylate |
Safety and Handling
| Hazard Category | Warning |