missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl (4R,6S)-6-(Hydroxymethyl)-2,2-dimethyl-1,3-dioxane-4-acetate 98.0+%, TCI America™

Catalog No. B42145G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42145G 5 g
B421425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B42145G Supplier TCI America Supplier No. B42145G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 124655-09-0
Molecular Formula C13H24O5
Molecular Weight (g/mol) 260.33
MDL Number MFCD23098985
InChI Key CFRUAOXMCVQMFP-ZJUUUORDSA-N
Synonym (4R,6S)-6-(Hydroxymethyl)-2,2-dimethyl-1,3-dioxane-4-acetic Acid tert-Butyl Ester, tert-Butyl 2-[(4R,6S)-6-(Hydroxymethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetate, 2-[(4R,6S)-6-(Hydroxymethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetic Acid tert-Butyl Ester
PubChem CID 9816650
IUPAC Name tert-butyl 2-[(4R,6S)-6-(hydroxymethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetate
SMILES CC1(OC(CC(O1)CO)CC(=O)OC(C)(C)C)C

Specifications

Color Yellow
Quantity 5 g
Formula Weight 260.33
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyl (4R,6S)-6-(Hydroxymethyl)-2,2-dimethyl-1,3-dioxane-4-acetate
EINECSNumber (5)-6817
TSCA No
Recommended Storage Refrigerator