missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-Formylbenzoate 98.0+%, TCI America™

Catalog No. b54141g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B54141G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B54141G Supplier TCI America Supplier No. B54141G

Chemical Identifiers

CAS 65874-27-3
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.241
MDL Number MFCD01111994
InChI Key DUNFNBQQWYQKFE-UHFFFAOYSA-N
Synonym 4-Formylbenzoic Acid tert-Butyl Ester, 4-(tert-Butoxycarbonyl)benzaldehyde
PubChem CID 2751588
IUPAC Name tert-butyl 4-formylbenzoate
SMILES CC(C)(C)OC(=O)C1=CC=C(C=C1)C=O

Specifications

Melting Point 54°C
Color White-Yellow
Quantity 1 g
Formula Weight 206.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material tert-Butyl 4-Formylbenzoate